accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 6006-82-2

IUPAC Name: 2-(2H-1,3-benzodioxol-5-yl)ethan-1-ol
Synonyms: 2-(2H-1,3-benzodioxol-5-yl)ethanol
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: JADSGOFBFPTCHG-UHFFFAOYSA-N
SMILES: OCCC1=CC2=C(OCO2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results