accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH3NH3H2OHOOOHO

CAS RN 6009-70-7

IUPAC Name: oxalic acid diamine hydrate
Synonyms: oxalic acid diamine hydrate
Molecular Weight (g/mol): 142.11
Molecular Formula: C2H10N2O5
InChi Key: MSMNVXKYCPHLLN-UHFFFAOYSA-N
SMILES: N.N.O.OC(=O)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (9)
  • (15)
Quantity 
  • (4)
  • (7)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (10)
Percent Purity 
  • (3)
  • (3)
  • (5)
  • (1)
  • (4)
  • (2)
Physical Form 
  • (2)
  • (6)
  • (3)
  • (3)
  • (4)
Grade 
  • (6)
  • (5)
  • (4)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (3)
  • (5)
  • (6)
  • (3)
  • (1)
  • (3)
  • (9)
  • (15)
  • (4)
  • (7)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (10)
  • (3)
  • (3)
  • (5)
  • (1)
  • (4)
  • (2)
  • (2)
  • (6)
  • (3)
  • (3)
  • (4)
  • (6)
  • (5)
  • (4)
  • (2)