accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3CH3OH3CH3C

CAS RN 601-57-0

IUPAC Name: 9a,11a-dimethyl-1-(6-methylheptan-2-yl)-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
Synonyms: 9a,11a-dimethyl-1-(6-methylheptan-2-yl)-1H,2H,3H,3aH,3bH,4H,5H,8H,9H,9bH,10H,11H-cyclopenta[a]phenanthren-7-one
Molecular Weight (g/mol): 384.65
Molecular Formula: C27H44O
InChi Key: NYOXRYYXRWJDKP-UHFFFAOYNA-N
SMILES: CC(C)CCCC(C)C1CCC2C3CCC4=CC(=O)CCC4(C)C3CCC12C
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results