accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 601-89-8

IUPAC Name: 2-nitrobenzene-1,3-diol
Synonyms: 2-nitroresorcinol
Molecular Weight (g/mol): 155.11
Molecular Formula: C6H5NO4
InChi Key: ZLCPKMIJYMHZMJ-UHFFFAOYSA-N
SMILES: OC1=CC=CC(O)=C1[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results