accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 60134-26-1

IUPAC Name: 2-(hydroxymethyl)-5-methoxyoxolan-3-ol
Synonyms: 2-(hydroxymethyl)-5-methoxyoxolan-3-ol
Molecular Weight (g/mol): 148.16
Molecular Formula: C6H12O4
InChi Key: NVGJZDFWPSOTHM-UHFFFAOYNA-N
SMILES: COC1CC(O)C(CO)O1
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results