CAS RN 60136-25-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
5-Isopropyl-2'-deoxyuridine, TRC
CAS: 60136-25-6 Molecular Formula: C12 H18 N2 O5 Molecular Weight (g/mol): 270.28 Synonym: 2'-Deoxy-5-(1-Methylethyl)Uridine,2'-Deoxy-5-Isopropyluridine,5-Isopropyldeoxyuridine,Epervudine SMILES: Cc(C)C1=Cn([C@H]2C[C@H](O)[C@@H](Co)O2)C(=O)Nc1=O