accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClFFFOOH

CAS RN 601513-31-9

IUPAC Name: 2-[4-chloro-2-(trifluoromethyl)phenyl]acetic acid
Synonyms: [4-chloro-2-(trifluoromethyl)phenyl]acetic acid
Molecular Weight (g/mol): 238.59
Molecular Formula: C9H6ClF3O2
InChi Key: UKSTVTZXWKFMHX-UHFFFAOYSA-N
SMILES: OC(=O)CC1=C(C=C(Cl)C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results