accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2OH2ONaNH2HOOO

CAS RN 6018-19-5

IUPAC Name: sodium 4-amino-2-hydroxybenzoate dihydrate
Synonyms: aminosalicylate sodium
Molecular Weight (g/mol): 211.15
Molecular Formula: C7H10NNaO5
InChi Key: GMUQJDAYXZXBOT-UHFFFAOYSA-M
SMILES: O.O.[Na+].NC1=CC=C(C([O-])=O)C(O)=C1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Physical Form 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results