accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHHO(?)

CAS RN 602-09-5

IUPAC Name: [1,1'-binaphthalene]-2,2'-diol
Synonyms: 1,1'-bi-2-naphthol
Molecular Weight (g/mol): 286.33
Molecular Formula: C20H14O2
InChi Key: PPTXVXKCQZKFBN-UHFFFAOYSA-N
SMILES: OC1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)