accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOOFFFFF

CAS RN 602-94-8

IUPAC Name: pentafluorobenzoic acid
Synonyms: pentafluorobenzoic acid sodium salt pentafluorobenzoic acid
Molecular Weight (g/mol): 212.08
Molecular Formula: C7HF5O2
InChi Key: YZERDTREOUSUHF-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(F)C(F)=C(F)C(F)=C1F
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results