accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 60230-37-7

IUPAC Name: 2,6-difluorobenzene-1-sulfonamide
Synonyms: 2,6-difluorobenzenesulfonamide
Molecular Weight (g/mol): 193.17
Molecular Formula: C6H5F2NO2S
InChi Key: RVVVGGCOFWWDEL-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C1=C(F)C=CC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results