accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 60239-18-1

HOONNNNHOOOHOOHO

CAS RN 60239-18-1

IUPAC Name: 2-[4,7,10-tris(carboxymethyl)-1,4,7,10-tetraazacyclododecan-1-yl]acetic acid
Synonyms: DOTA dota (chelator) CuDOTA tetraxetan dota acid
Molecular Weight (g/mol): 404.42
Molecular Formula: C16H28N4O8
InChi Key: WDLRUFUQRNWCPK-UHFFFAOYSA-N
SMILES: OC(=O)CN1CCN(CC(O)=O)CCN(CC(O)=O)CCN(CC(O)=O)CC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results