accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 60282-87-3

CH3O(R)HOHC

CAS RN 60282-87-3

IUPAC Name: (1R)-11a-ethyl-1-ethynyl-1-hydroxy-1H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
Synonyms: gestodene
Molecular Weight (g/mol): 310.44
Molecular Formula: C21H26O2
InChi Key: SIGSPDASOTUPFS-KQMXEUTGSA-N
SMILES: CCC12CCC3C(CCC4=CC(=O)CCC34)C1C=C[C@@]2(O)C#C
Molecular Weight (g/mol) 
  • (2)
  • (2)
Certifications/Compliance 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)