CAS RN 603-44-1
IUPAC Name:
4-[bis(4-hydroxyphenyl)methyl]phenol
Synonyms:
4-[bis(4-hydroxyphenyl)methyl]phenol
Molecular Weight (g/mol):
292.33
Molecular Formula:
C19H16O3
InChi Key:
WFCQTAXSWSWIHS-UHFFFAOYSA-N
SMILES:
OC1=CC=C(C=C1)C(C1=CC=C(O)C=C1)C1=CC=C(O)C=C1
1
–
1
of
1
results
1
4,4',4″-Trihydroxytriphenylmethane, 97%, Thermo Scientific™
CAS: 603-44-1 Molecular Formula: C19H16O3 Molecular Weight (g/mol): 292.33 MDL Number: MFCD00191589 InChI Key: WFCQTAXSWSWIHS-UHFFFAOYSA-N Synonym: 4,4',4-methanetriyltriphenol,leucoaurin,4,4',4-trihydroxytriphenylmethane,leucoaurine,leucaurin,phenol, methylidynetris,4-bis 4-hydroxyphenyl methyl phenol,4,4',4-methylidynetriphenol,4,4',4-methylidynetrisphenol,phenol, 4,4',4-methylidynetris PubChem CID: 69047 IUPAC Name: 4-[bis(4-hydroxyphenyl)methyl]phenol SMILES: OC1=CC=C(C=C1)C(C1=CC=C(O)C=C1)C1=CC=C(O)C=C1