accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CH3H3CNOO

CAS RN 603-71-4

IUPAC Name: 1,3,5-trimethyl-2-nitrobenzene
Synonyms: 1,3,5-trimethyl-2-nitrobenzene
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: SCEKDQTVGHRSNS-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(C(C)=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results