accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3ClNOO

CAS RN 60468-54-4

IUPAC Name: 1-(chloromethyl)-2-methyl-3-nitrobenzene
Synonyms: 1-(chloromethyl)-2-methyl-3-nitrobenzene
Molecular Weight (g/mol): 185.61
Molecular Formula: C8H8ClNO2
InChi Key: XZNDXQGZPOZITR-UHFFFAOYSA-N
SMILES: CC1=C(CCl)C=CC=C1[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results