accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 605-02-7

CAS RN 605-02-7

IUPAC Name: 1-phenylnaphthalene
Synonyms: naphthalene, 1-phenyl-
Molecular Weight (g/mol): 204.27
Molecular Formula: C16H12
InChi Key: IYDMICQAKLQHLA-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)C1=C2C=CC=CC2=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results