accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 605-48-1

IUPAC Name: 9,10-dichloroanthracene
Synonyms: 9,10-dichloroanthracene
Molecular Weight (g/mol): 247.12
Molecular Formula: C14H8Cl2
InChi Key: FKDIWXZNKAZCBY-UHFFFAOYSA-N
SMILES: ClC1=C2C=CC=CC2=C(Cl)C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results