accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ICH3NNCH3(E/Z)(E/Z)

CAS RN 605-91-4

IUPAC Name: 1-ethyl-2-[3-(1-ethyl-1,2-dihydroquinolin-2-ylidene)prop-1-en-1-yl]quinolin-1-ium iodide
Synonyms: carbocyanine pinacyanol iodide pinacyanole sensitol red
Molecular Weight (g/mol): 480.39
Molecular Formula: C25H25IN2
InChi Key: QWYZFXLSWMXLDM-UHFFFAOYSA-M
SMILES: [I-].CCN1C(=CC=CC2=CC=C3C=CC=CC3=[N+]2CC)C=CC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results