accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2NOHONOOOHO

CAS RN 606-59-7

IUPAC Name: 2-amino-3-oxo-3H-phenoxazine-1,9-dicarboxylic acid
Synonyms: cinnabarinic acid cinnavalininate
Molecular Weight (g/mol): 300.23
Molecular Formula: C14H8N2O6
InChi Key: FSBKJYLVDRVPTK-UHFFFAOYSA-N
SMILES: NC1=C(C(O)=O)C2=NC3=C(C=CC=C3OC2=CC1=O)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results