accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

TaOOOOOH3CCH3H3CCH3CH3

CAS RN 6074-84-6

IUPAC Name: tantalum(5+) pentaethanolate
Synonyms: tantalum(V) ethoxide
Molecular Weight (g/mol): 406.25
Molecular Formula: C10H25O5Ta
InChi Key: HSXKFDGTKKAEHL-UHFFFAOYSA-N
SMILES: [Ta+5].CC[O-].CC[O-].CC[O-].CC[O-].CC[O-]
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results