accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 608-31-1

IUPAC Name: 2,6-dichloroaniline
Synonyms: 2,6-dichloroaniline
Molecular Weight (g/mol): 162.01
Molecular Formula: C6H5Cl2N
InChi Key: JDMFXJULNGEPOI-UHFFFAOYSA-N
SMILES: NC1=C(Cl)C=CC=C1Cl
Molecular Weight (g/mol) 
  • (6)
  • (5)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results