accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2OH2OH2OPbCH3OOCH3OO

CAS RN 6080-56-4

IUPAC Name: λ²-lead(2+) diacetate trihydrate
Synonyms: λ²-lead(2+) diacetate trihydrate
Molecular Weight (g/mol): 379.3
Molecular Formula: C4H12O7Pb
InChi Key: MCEUZMYFCCOOQO-UHFFFAOYSA-L
SMILES: O.O.O.[Pb++].CC([O-])=O.CC([O-])=O
Molecular Weight (g/mol) 
  • (34)
  • (2)
Quantity 
  • (4)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (4)
  • (1)
  • (5)
  • (3)
  • (1)
  • (1)
  • (1)
  • (6)
Percent Purity 
  • (7)
  • (3)
  • (1)
  • (2)
  • (6)
  • (3)
  • (3)
  • (2)
Grade 
  • (3)
  • (7)
  • (3)
  • (3)
  • (1)
  • (3)
  • (2)
  • (4)
Physical Form 
  • (3)
  • (9)
  • (3)
  • (9)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (6)
  • (10)
  • (1)
  • (9)
  • (8)
  • (2)
  • (1)
  • (34)
  • (2)
  • (4)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (4)
  • (1)
  • (5)
  • (3)
  • (1)
  • (1)
  • (1)
  • (6)
  • (7)
  • (3)
  • (1)
  • (2)
  • (6)
  • (3)
  • (3)
  • (2)
  • (3)
  • (7)
  • (3)
  • (3)
  • (1)
  • (3)
  • (2)
  • (4)
  • (3)
  • (9)
  • (3)
  • (9)