accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2BrCl

CAS RN 60811-17-8

IUPAC Name: 5-bromo-2-chloroaniline
Synonyms: 5-bromo-2-chloroaniline
Molecular Weight (g/mol): 206.47
Molecular Formula: C6H5BrClN
InChi Key: UGOLEPGQWYPIBR-UHFFFAOYSA-N
SMILES: NC1=CC(Br)=CC=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results