accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 60832-72-6

IUPAC Name: 2H,3H-[1,3]oxazolo[4,5-b]pyridin-2-one
Synonyms: 3H-[1,3]oxazolo[4,5-b]pyridin-2-one
Molecular Weight (g/mol): 136.11
Molecular Formula: C6H4N2O2
InChi Key: OVLXOTUWFLHWQT-UHFFFAOYSA-N
SMILES: O=C1NC2=NC=CC=C2O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results