CAS RN 608534-41-4
CAS RN 608534-41-4
IUPAC Name:
9-[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)ethyl]-9H-carbazole
Synonyms:
9-[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)ethyl]carbazole
Molecular Weight (g/mol):
321.23
Molecular Formula:
C20H24BNO2
InChi Key:
NTUZDLWSQJRXAM-UHFFFAOYSA-N
SMILES:
CC1(C)OB(CCN2C3=C(C=CC=C3)C3=C2C=CC=C3)OC1(C)C
1
–
1
of
1
results
1
2-(9-Carbazolyl)ethylboronic acid pinacol ester, 98%
CAS: 608534-41-4 Molecular Formula: C20H24BNO2 Molecular Weight (g/mol): 321.227 MDL Number: MFCD03788738 InChI Key: NTUZDLWSQJRXAM-UHFFFAOYSA-N Synonym: 2-9-carbazolyl ethylboronic acid pinacol ester,9-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl ethyl-9h-carbazole,9-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl ethyl carbazole,9-2-tetramethyl-1,3,2-dioxaborolan-2-yl ethyl carbazole,2-2-9h-carbazole-9-yl ethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane PubChem CID: 45925651 IUPAC Name: 9-[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)ethyl]carbazole SMILES: B1(OC(C(O1)(C)C)(C)C)CCN2C3=CC=CC=C3C4=CC=CC=C42