accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NNN

CAS RN 6086-21-1

IUPAC Name: 1-methyl-1H-1,2,4-triazole
Synonyms: 1-methyl-1,2,4-triazole
Molecular Weight (g/mol): 83.09
Molecular Formula: C3H5N3
InChi Key: MWZDIEIXRBWPLG-UHFFFAOYSA-N
SMILES: CN1C=NC=N1
Quantity 
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results