accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 6088-88-6

H3CMgCl(E)

CAS RN 6088-88-6

IUPAC Name: (2E)-3-(4-chlorophenyl)-2-[(furan-2-yl)formamido]prop-2-enamide
Synonyms: (2E)-3-(4-chlorophenyl)-2-(furan-2-ylformamido)prop-2-enamide
Molecular Weight (g/mol): 290.7
Molecular Formula: C14H11ClN2O3
InChi Key: REELYLPJMWRIQG-DHZHZOJOSA-N
SMILES: NC(=O)C(\NC(=O)C1=CC=CO1)=C/C1=CC=C(Cl)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (1)