accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CHOO

CAS RN 6089-04-9

IUPAC Name: 2-(prop-2-yn-1-yloxy)oxane
Synonyms: 2-(prop-2-yn-1-yloxy)oxane
Molecular Weight (g/mol): 140.18
Molecular Formula: C8H12O2
InChi Key: HQAXHIGPGBPPFU-UHFFFAOYNA-N
SMILES: C#CCOC1CCCCO1
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)