CAS RN 609-39-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Nitrofuran, TRC
CAS: 609-39-2 Molecular Formula: C4H3NO3 Molecular Weight (g/mol): 113.07 Synonym: NSC 59983,Nitrofuran IUPAC Name: 2-nitrofuran SMILES: [O-][N+](=O)c1occc1