accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 609-54-1

CH3CH3SHOOO

CAS RN 609-54-1

IUPAC Name: 2,5-dimethylbenzene-1-sulfonic acid
Synonyms: 2,5-dimethylbenzenesulfonic acid
Molecular Weight (g/mol): 186.23
Molecular Formula: C8H10O3S
InChi Key: IRLYGRLEBKCYPY-UHFFFAOYSA-N
SMILES: CC1=CC=C(C)C(=C1)S(O)(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results