accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 609-85-8

IUPAC Name: 2-amino-3,5-dibromobenzoic acid
Synonyms: 2-amino-3,5-dibromobenzoic acid
Molecular Weight (g/mol): 294.93
Molecular Formula: C7H5Br2NO2
InChi Key: WNABMWFLKQEGCP-UHFFFAOYSA-N
SMILES: NC1=C(Br)C=C(Br)C=C1C(O)=O
Quantity 
  • (2)
  • (2)
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results