accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 60933-65-5

CH3SHNO

CAS RN 60933-65-5

IUPAC Name: imino(methyl)phenyl-λ⁶-sulfanone
Synonyms: imino(methyl)phenyl-λ⁶-sulfanone
Molecular Weight (g/mol): 155.22
Molecular Formula: C7H9NOS
InChi Key: YFYIDTVGWCYSEO-UHFFFAOYNA-N
SMILES: CS(=N)(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results