accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 60966-26-9

IUPAC Name: 2-amino-9-[3-amino-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-3H-purin-6-one
Synonyms: 2-amino-9-[3-amino-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
Molecular Weight (g/mol): 282.26
Molecular Formula: C10H14N6O4
InChi Key: ROPTVRLUGSPXNH-UHFFFAOYNA-N
SMILES: NC1C(O)C(CO)OC1N1C=NC2=C1NC(N)=NC2=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results