accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 61-80-3

IUPAC Name: 5-chloro-1,3-benzoxazol-2-amine
Synonyms: flexin zoxazolamine (inn) zoxazolamine 氯苯唑胺
Molecular Weight (g/mol): 168.58
Molecular Formula: C7H5ClN2O
InChi Key: YGCODSQDUUUKIV-UHFFFAOYSA-N
SMILES: NC1=NC2=CC(Cl)=CC=C2O1
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results