CAS RN 610-22-0
IUPAC Name:
1,2-dimethyl 4-nitrobenzene-1,2-dicarboxylate
Synonyms:
1,2-dimethyl 4-nitrophthalate
Molecular Weight (g/mol):
239.18
Molecular Formula:
C10H9NO6
InChi Key:
XWBDWELWBUWSNI-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC=C(C=C1C(=O)OC)[N+]([O-])=O
1
–
3
of
3
results
2
4-Nitrophthalic acid, bis-methyl ester 100 μg/mL in Hexane, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
3
Dimethyl 4-nitrophthalate, 98%, Thermo Scientific™
CAS: 610-22-0 Molecular Formula: C10H9NO6 Molecular Weight (g/mol): 239.18 MDL Number: MFCD00017191 InChI Key: XWBDWELWBUWSNI-UHFFFAOYSA-N PubChem CID: 69119 IUPAC Name: dimethyl 4-nitrobenzene-1,2-dicarboxylate SMILES: COC(=O)C1=CC=C(C=C1C(=O)OC)[N+]([O-])=O