accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 610-37-7

IUPAC Name: 5-hydroxy-2-nitrobenzoic acid
Synonyms: 5-hydroxy-2-nitrobenzoic acid
Molecular Weight (g/mol): 183.12
Molecular Formula: C7H5NO5
InChi Key: BUHKQTKKZAXSMH-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(O)=CC=C1[N+]([O-])=O
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results