CAS RN 61024-31-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Chloro Fenofibric Acid, TRC
CAS: 61024-31-5 Molecular Formula: C17H15ClO4 Molecular Weight (g/mol): 318.75 Synonym: 2-[4-(2-Chlorobenzoyl)phenoxy]-2-methylpropanoic Acid,LF 590 IUPAC Name: 2-[4-(2-chlorobenzoyl)phenoxy]-2-methylpropanoic acid SMILES: CC(C)(Oc1ccc(cc1)C(=O)c2ccccc2Cl)C(=O)O