accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OSOOCl

CAS RN 610277-83-3

IUPAC Name: 4-(2-methylphenoxy)benzene-1-sulfonyl chloride
Synonyms: 4-(2-methylphenoxy)benzenesulfonyl chloride
Molecular Weight (g/mol): 282.74
Molecular Formula: C13H11ClO3S
InChi Key: IJPKGMOVNZHZKZ-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1OC1=CC=C(C=C1)S(Cl)(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results