accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 610768-32-6

BOOH3CCH3H3CH3CNCH3

CAS RN 610768-32-6

IUPAC Name: 2-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Synonyms: 2-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Molecular Weight (g/mol): 219.09
Molecular Formula: C12H18BNO2
InChi Key: JEECGGXGHJUYMN-UHFFFAOYSA-N
SMILES: CC1=NC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results