accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 6108-17-4

IUPAC Name: lithium(1+) acetate dihydrate
Synonyms: lithium(1+) acetate dihydrate
Molecular Weight (g/mol): 102.01
Molecular Formula: C2H7LiO4
InChi Key: IAQLJCYTGRMXMA-UHFFFAOYSA-M
SMILES: [Li+].O.O.CC([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (7)
  • (4)
Quantity 
  • (5)
  • (1)
  • (7)
Physical Form 
  • (2)
  • (2)
  • (1)
  • (4)
Grade 
  • (2)
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (4)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (2)
  • (7)
  • (4)
  • (5)
  • (1)
  • (7)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)