accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

LiLiOOOHOOOHH2O

CAS RN 6108-32-3

IUPAC Name: dilithium(1+) 2,3-dihydroxybutanedioate hydrate
Synonyms: dilithium(1+) hydrate tartrate
Molecular Weight (g/mol): 179.97
Molecular Formula: C4H6Li2O7
InChi Key: GNDOSKKVQUGWFB-UHFFFAOYNA-L
SMILES: [Li+].[Li+].O.OC(C(O)C([O-])=O)C([O-])=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results