accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NH2NOO

CAS RN 611-05-2

IUPAC Name: 3-methyl-4-nitroaniline
Synonyms: 3-methyl-4-nitroaniline
Molecular Weight (g/mol): 152.15
Molecular Formula: C7H8N2O2
InChi Key: XPAYEWBTLKOEDA-UHFFFAOYSA-N
SMILES: CC1=CC(N)=CC=C1[N+]([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results