accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2N

CAS RN 611-34-7

IUPAC Name: quinolin-5-amine
Synonyms: 5-aminoquinoline
Molecular Weight (g/mol): 144.18
Molecular Formula: C9H8N2
InChi Key: XMIAFAKRAAMSGX-UHFFFAOYSA-N
SMILES: NC1=C2C=CC=NC2=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (3)
  • (1)
  • (3)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (3)
  • (4)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)