accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHN

CAS RN 611-36-9

IUPAC Name: 1,4-dihydroquinolin-4-one
Synonyms: 4(1H)-quinolinone kynurine
Molecular Weight (g/mol): 145.16
Molecular Formula: C9H7NO
InChi Key: PMZDQRJGMBOQBF-UHFFFAOYSA-N
SMILES: O=C1C=CNC2=CC=CC=C12
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)