accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 611-53-0

IUPAC Name: 4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodo-1,2-dihydropyrimidin-2-one
Synonyms: 5-iodo-2'-deoxycytidine ibacitabine
Molecular Weight (g/mol): 353.12
Molecular Formula: C9H12IN3O4
InChi Key: WEVJJMPVVFNAHZ-RRKCRQDMSA-N
SMILES: NC1=NC(=O)N(C=C1I)[C@H]1C[C@H](O)[C@@H](CO)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results