CAS RN 61196-37-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
rac-Praziquanamine, TRC
CAS: 61196-37-0 Molecular Formula: C12 H14 N2 O Molecular Weight (g/mol): 202.2524 Synonym: 1,2,3,6,7,11b-Hexahydro-4H-pyrazino[2,1-a]isoquinolin-4-one,(±)-Praziquanamine,DL-Praziquanamine IUPAC Name: 1,2,3,6,7,11b-hexahydropyrazino[2,1-a]isoquinolin-4-one SMILES: O=C1CNCC2N1CCc3ccccc23