accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 612-05-5

IUPAC Name: 2-methoxyoxane-3,4,5-triol
Synonyms: 2-methoxyoxane-3,4,5-triol
Molecular Weight (g/mol): 164.16
Molecular Formula: C6H12O5
InChi Key: ZBDGHWFPLXXWRD-UHFFFAOYNA-N
SMILES: COC1OCC(O)C(O)C1O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results