accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 612-82-8

IUPAC Name: dihydrogen 3,3'-dimethyl-[1,1'-biphenyl]-4,4'-diamine dichloride
Synonyms: dihydrogen tolidine dichloride
Molecular Weight (g/mol): 285.21
Molecular Formula: C14H18Cl2N2
InChi Key: LUKPNZHXJRJBAN-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].CC1=CC(=CC=C1N)C1=CC=C(N)C(C)=C1
Molecular Weight (g/mol) 
  • (10)
  • (5)
Quantity 
  • (4)
  • (1)
  • (1)
  • (4)
  • (1)
  • (3)
  • (1)
Physical Form 
  • (4)
  • (3)
Grade 
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (5)
  • (3)
  • (10)
  • (5)
  • (4)
  • (1)
  • (1)
  • (4)
  • (1)
  • (3)
  • (1)
  • (4)
  • (3)
  • (3)
  • (3)
  • (2)
  • (4)