accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ON

CAS RN 613-90-1

IUPAC Name: benzoyl cyanide
Synonyms: benzoyl cyanide α-oxobenzeneacetonitrile benzoyl nitrile α-oxo-α-tolunitrile
Molecular Weight (g/mol): 131.13
Molecular Formula: C8H5NO
InChi Key: GJQBHOAJJGIPRH-UHFFFAOYSA-N
SMILES: O=C(C#N)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results